Zestawy testowe, fotometryczne, aluminium, testy kuwetowe LCK
Catalog # 705-3003
Supplier: Hach
CAS Number:
Specifications
- Metoda chemiczna:Chromazurol S
- Ilość:24 Tests
- Opis:Glin
- Seq:203
- Typ:LCK301
- Dla urządzeń:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Zakres:0,02 - 0,5 mg/l Al
Specifications
About this item
Ready for use, photometric evaluation system and environmentally friendly.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C