Uszczelka samoprzylepna, AMPLIseal
736-0354
- Właściwość optyczna:Transparent
- Opis:AMPLIseal™
- Ilość:100
AMPLIseal™ is a self-adhesive sealing film notable for its low autofluorescence, in particular in the wavelength range critical for Real Time PCR. It consists of a thick PP film coated with a transparent acrylate adhesive. The adhesive layer is protected by a peelable polyester film. The strongly adhering acrylate adhesive provides reliable sealing, thereby minimising evaporation but without influencing the PCR reaction or the fluorescence measurement.
- Highly transparent with minimal autofluorescence
- Ideal for Real Time PCR