Order Entry
Poland
ContactUsLinkComponent
Pseudomonas aeruginosa WDCM 00024 Vitroids 1,000-10,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000246-10EA
Supplier:  Merck
New Product
Pseudomonas aeruginosa WDCM 00024 Vitroids 1,000-10,000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000246-10EA
Supplier:  Merck