Order Entry
Poland
ContactUsLinkComponent
Thermo Fisher Scientific Refrigerated Incubator RI 150, Chamber Volume 150L, 4 to 60°C, Solid Door, 230V 1 * 1 items
numer katalogowy THBRTFSRI150-230V
New Product
Thermo Fisher Scientific Refrigerated Incubator RI 150, Chamber Volume 150L, 4 to 60°C, Solid Door, 230V 1 * 1 items
numer katalogowy THBRTFSRI150-230V