Order Entry
Poland
ContactUsLinkComponent
Mouse Syntenin 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Syntenin 2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA270930-48
New Product
Mouse Syntenin 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Syntenin 2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA270930-48