Order Entry
Poland
ContactUsLinkComponent
Rat Ghrelin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinati on of rat Ghrelin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flu 1 * 48 Tests
Catalog # ANTIA2419-48
Supplier:  ANTIBODIES.COM
New Product
Rat Ghrelin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinati on of rat Ghrelin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flu 1 * 48 Tests
Catalog # ANTIA2419-48
Supplier:  ANTIBODIES.COM