Order Entry
Poland
ContactUsLinkComponent
Mouse BMP1/PCP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse BMP1/PCP in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2375-48
Supplier:  ANTIBODIES.COM
New Product
Mouse BMP1/PCP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse BMP1/PCP in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2375-48
Supplier:  ANTIBODIES.COM