Order Entry
Poland
ContactUsLinkComponent
Human ALDH7A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human ALDH7A1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA270639-48
Supplier:  Antibodies.com
New Product
Human ALDH7A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human ALDH7A1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA270639-48
Supplier:  Antibodies.com