Order Entry
Poland
ContactUsLinkComponent
Human Agrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Agrin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA2324-48
New Product
Human Agrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Agrin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA2324-48