Human Natriuretic Peptides A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human Natriuretic Peptides A in serum, plasma, tissue homogenates, cell lysates, cell culture sup 1 * 48 Tests
Catalog # ANTIA2700-48
Supplier: Antibodies.com