Order Entry
Poland
ContactUsLinkComponent
Human GilZ/TilZ ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human GilZ/TilZ in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2421-48
Supplier:  Antibodies.com
New Product
Human GilZ/TilZ ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human GilZ/TilZ in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2421-48
Supplier:  Antibodies.com