Order Entry
Poland
ContactUsLinkComponent
Rat Granzyme B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat Granzyme B in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2029-48
Supplier:  Antibodies.com
New Product
Rat Granzyme B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat Granzyme B in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2029-48
Supplier:  Antibodies.com