Order Entry
Poland
ContactUsLinkComponent
Human NEU2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human NEU2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA270792-48
New Product
Human NEU2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human NEU2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA270792-48