Order Entry
Poland
ContactUsLinkComponent
Human Presenilin 2/AD5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Presenilin 2/AD5 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270647-48
Supplier:  Antibodies.com
New Product
Human Presenilin 2/AD5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Presenilin 2/AD5 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270647-48
Supplier:  Antibodies.com