Order Entry
Poland
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Dopamine Receptor D3/DRD3 protein to be purified and analysed in vitro. 1 * 50 µG
numer katalogowy ANTIA325119-50
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Dopamine Receptor D3/DRD3 protein to be purified and analysed in vitro. 1 * 50 µG
numer katalogowy ANTIA325119-50
Supplier:  ANTIBODIES.COM