Order Entry
Poland
ContactUsLinkComponent
Consists of amino acids Arg319-Phe541 of SARS-CoV-2 Spike Glycoprotein RBD (Kappa B.1.617/B.1.617.1/B.1.617.3 Variant) containing th e following mutations: L452R and E484Q. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA318168-50
Supplier:  Antibodies.com
New Product
Consists of amino acids Arg319-Phe541 of SARS-CoV-2 Spike Glycoprotein RBD (Kappa B.1.617/B.1.617.1/B.1.617.3 Variant) containing th e following mutations: L452R and E484Q. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA318168-50
Supplier:  Antibodies.com