Order Entry
Poland
ContactUsLinkComponent
Human Leptin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Leptin in serum, plasma, saliva, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 48 Tests
  ANTIA2988-48
New Product
Human Leptin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Leptin in serum, plasma, saliva, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 48 Tests
  ANTIA2988-48