Order Entry
Poland
ContactUsLinkComponent
Human Vitamin D Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Vitamin D Receptor in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3207-48
Supplier:  Antibodies.com
New Product
Human Vitamin D Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Vitamin D Receptor in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3207-48
Supplier:  Antibodies.com