Order Entry
Poland
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Claudin 7/CLDN-7 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA317340-10
Supplier:  Antibodies.com
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Claudin 7/CLDN-7 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA317340-10
Supplier:  Antibodies.com