Order Entry
Poland
ContactUsLinkComponent
Rat Brevican ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Brevican in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA326114-48
Supplier:  Antibodies.com
New Product
Rat Brevican ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Brevican in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA326114-48
Supplier:  Antibodies.com