Order Entry
Poland
ContactUsLinkComponent
Human NGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
  ANTIA3032-48
New Product
Human NGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
  ANTIA3032-48