Order Entry
Poland
ContactUsLinkComponent
Mouse CD8 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse CD8 alpha in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA326124-48
Supplier:  Antibodies.com
New Product
Mouse CD8 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse CD8 alpha in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA326124-48
Supplier:  Antibodies.com