Order Entry
Poland
ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Glucagon Receptor protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA318426-10
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Glucagon Receptor protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA318426-10
Supplier:  ANTIBODIES.COM