Order Entry
Poland
ContactUsLinkComponent
Androstenedione ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of Androstenedione in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 48 Tests
Catalog # ANTIA3192-48
Supplier:  Antibodies.com
New Product
Androstenedione ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of Androstenedione in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 48 Tests
Catalog # ANTIA3192-48
Supplier:  Antibodies.com