Order Entry
Poland
ContactUsLinkComponent
Human VEGF Receptor 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human VEGF Receptor 3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2932-48
Supplier:  Antibodies.com
New Product
Human VEGF Receptor 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human VEGF Receptor 3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2932-48
Supplier:  Antibodies.com