Order Entry
Poland
ContactUsLinkComponent
Porcine GHRH ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinat ion of porcine GHRH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological f 1 * 48 Tests
Catalog # ANTIA3241-48
Supplier:  Antibodies.com
New Product
Porcine GHRH ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinat ion of porcine GHRH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological f 1 * 48 Tests
Catalog # ANTIA3241-48
Supplier:  Antibodies.com