Order Entry
Poland
ContactUsLinkComponent
Porcine Tissue Plasminogen Activator ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of porcine Tissue Plasminogen Activator in plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3222-48
Supplier:  Antibodies.com
New Product
Porcine Tissue Plasminogen Activator ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of porcine Tissue Plasminogen Activator in plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3222-48
Supplier:  Antibodies.com