Order Entry
Poland
ContactUsLinkComponent
Rat Luteinizing Hormone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Luteinizing Hormone in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3153-48
Supplier:  Antibodies.com
New Product
Rat Luteinizing Hormone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of rat Luteinizing Hormone in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3153-48
Supplier:  Antibodies.com