Order Entry
Poland
ContactUsLinkComponent
Human Epstein Barr Virus EA-D IgM ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determ ination of human EA-D EBV IgM in serum. 1 * 96 Tests
Catalog # ANTIA324355-96
Supplier:  ANTIBODIES.COM
New Product
Human Epstein Barr Virus EA-D IgM ELISA Kit is an Enzyme-Linked Immunsorbent Assay designed for the in vitro semiquantitative determ ination of human EA-D EBV IgM in serum. 1 * 96 Tests
Catalog # ANTIA324355-96
Supplier:  ANTIBODIES.COM