Order Entry
Poland
ContactUsLinkComponent
Human Glutaminyl Cyclase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Glutaminyl Cyclase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5392-48
Supplier:  Antibodies.com
New Product
Human Glutaminyl Cyclase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Glutaminyl Cyclase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5392-48
Supplier:  Antibodies.com