Order Entry
Poland
ContactUsLinkComponent
Human MAPRE1/EB1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human MAPRE1/EB1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA4848-48
New Product
Human MAPRE1/EB1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human MAPRE1/EB1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA4848-48