Order Entry
Poland
ContactUsLinkComponent
Human CD59 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human CD59 in serum, plasma, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4916-48
Supplier:  Antibodies.com
New Product
Human CD59 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human CD59 in serum, plasma, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4916-48
Supplier:  Antibodies.com