Order Entry
Poland
ContactUsLinkComponent
Porcine Complement C3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of porcine Complement C3 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3290-48
Supplier:  Antibodies.com
New Product
Porcine Complement C3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of porcine Complement C3 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3290-48
Supplier:  Antibodies.com