Order Entry
Poland
ContactUsLinkComponent
Mouse NADPH Oxidase 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse NADPH Oxidase 4 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3882-48
Supplier:  Antibodies.com
New Product
Mouse NADPH Oxidase 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse NADPH Oxidase 4 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3882-48
Supplier:  Antibodies.com