Order Entry
Poland
ContactUsLinkComponent
Human FSH-R ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinati on of human FSH-R in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5448-96
Supplier:  ANTIBODIES.COM
New Product
Human FSH-R ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determinati on of human FSH-R in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5448-96
Supplier:  ANTIBODIES.COM