Order Entry
Poland
ContactUsLinkComponent
Rat Alanine Transaminase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Alanine Transaminase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA3701-48
Supplier:  Antibodies.com
New Product
Rat Alanine Transaminase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Alanine Transaminase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA3701-48
Supplier:  Antibodies.com