Order Entry
Poland
ContactUsLinkComponent
Human AANAT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AANAT in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3707-48
Supplier:  Antibodies.com
New Product
Human AANAT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AANAT in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3707-48
Supplier:  Antibodies.com