Order Entry
Poland
ContactUsLinkComponent
Human Topoisomerase I ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Topoisomerase I in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4811-96
Supplier:  ANTIBODIES.COM
New Product
Human Topoisomerase I ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Topoisomerase I in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4811-96
Supplier:  ANTIBODIES.COM