Order Entry
Poland
ContactUsLinkComponent
Rat Fibrinogen alpha Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Fibrinogen alpha Chain in plasma. 1 * 96 Tests
Catalog # ANTIA3661-96
Supplier:  ANTIBODIES.COM
New Product
Rat Fibrinogen alpha Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Fibrinogen alpha Chain in plasma. 1 * 96 Tests
Catalog # ANTIA3661-96
Supplier:  ANTIBODIES.COM