Order Entry
Poland
ContactUsLinkComponent
Chicken FGF4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of chicken FGF4 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA4570-96
New Product
Chicken FGF4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of chicken FGF4 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA4570-96