Order Entry
Poland
ContactUsLinkComponent
Human Nucleoporin p62/NUP62 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Nucleoporin p62/NUP62 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5398-96
Supplier:  ANTIBODIES.COM
New Product
Human Nucleoporin p62/NUP62 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Nucleoporin p62/NUP62 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5398-96
Supplier:  ANTIBODIES.COM