Order Entry
Poland
ContactUsLinkComponent
Human CYP24A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human CYP24A1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5192-96
Supplier:  ANTIBODIES.COM
New Product
Human CYP24A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human CYP24A1 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5192-96
Supplier:  ANTIBODIES.COM