Order Entry
Poland
ContactUsLinkComponent
Human PXR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PXR in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5143-96
Supplier:  ANTIBODIES.COM
New Product
Human PXR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PXR in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5143-96
Supplier:  ANTIBODIES.COM