Order Entry
Poland
ContactUsLinkComponent
Human Keratocan/KTN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Keratocan/KTN in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5500-96
Supplier:  ANTIBODIES.COM
New Product
Human Keratocan/KTN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Keratocan/KTN in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5500-96
Supplier:  ANTIBODIES.COM