Order Entry
Poland
ContactUsLinkComponent
Mouse GDF3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse GDF3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5578-96
Supplier:  ANTIBODIES.COM
New Product
Mouse GDF3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse GDF3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5578-96
Supplier:  ANTIBODIES.COM