Order Entry
Poland
ContactUsLinkComponent
Rat Aromatase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat Aromatase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4986-96
Supplier:  ANTIBODIES.COM
New Product
Rat Aromatase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat Aromatase in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4986-96
Supplier:  ANTIBODIES.COM