Order Entry
Poland
ContactUsLinkComponent
Human Glucagon Like Peptide 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of human Glucagon Like Peptide 2 in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 48 Tests
Catalog # ANTIA4918-48
Supplier:  Antibodies.com
New Product
Human Glucagon Like Peptide 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of human Glucagon Like Peptide 2 in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 48 Tests
Catalog # ANTIA4918-48
Supplier:  Antibodies.com