Order Entry
Poland
ContactUsLinkComponent
Bovine NGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f bovine NGF in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3943-96
Supplier:  ANTIBODIES.COM
New Product
Bovine NGF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f bovine NGF in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3943-96
Supplier:  ANTIBODIES.COM