Order Entry
Poland
ContactUsLinkComponent
Human Hepcidin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Hepcidin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA3770-96
Supplier:  ANTIBODIES.COM
New Product
Human Hepcidin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Hepcidin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA3770-96
Supplier:  ANTIBODIES.COM