Order Entry
Poland
ContactUsLinkComponent
Rat Lipin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Lipin 1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5960-48
Supplier:  ANTIBODIES.COM
New Product
Rat Lipin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Lipin 1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5960-48
Supplier:  ANTIBODIES.COM